Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426825.1 | XP_022282533.1 | 6 - 7 | 3324 | 60 | |
XM_022426826.1 | XP_022282534.1 | 6 - 7 | 3358 | 60 | ||
XM_022426827.1 | XP_022282535.1 | 6 - 7 | 1908 | 60 | ||
XM_022426828.1 | XP_022282536.1 | 5 - 6 | 3620 | 60 | ||
XM_022426829.1 | XP_022282537.1 | 5 - 6 | 3661 | 60 | ||
XM_022426830.1 | XP_022282538.1 | 4 - 5 | 3510 | 60 | ||
XM_022426831.1 | XP_022282539.1 | 5 - 6 | 3665 | 60 | ||
XM_539192.5 | XP_539192.3 | 5 - 6 | 3665 | 60 | ||
GenBank mRNA | DQ195094.1 | - | 3 - 4 | 543 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Inventoried Probe spans exons |