Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197043.1 | NP_001183972.1 | 3 - 4 | 544 | 79 | |
XM_005623184.3 | XP_005623241.1 | 4 - 5 | 680 | 79 | ||
GenBank mRNA | HQ637386.1 | - | 3 - 4 | 539 | 79 | |
L23429.1 | - | 2 - 3 | 533 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |