Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003004.1 | NP_001003004.1 | 9 - 10 | 1286 | 72 | |
XM_005636669.3 | XP_005636726.1 | 9 - 10 | 1997 | 72 | ||
GenBank mRNA | DQ672621.1 | - | 9 - 10 | 1287 | 72 | |
DQ672622.1 | - | 8 - 9 | 1563 | 72 | ||
EU285563.1 | - | 9 - 10 | 1287 | 72 | ||
U52916.1 | - | 9 - 10 | 1286 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |