Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003287.1 | NP_001003287.1 | 1 - 2 | 140 | 80 | |
GenBank mRNA | X53564.1 | - | 85 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |