Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010943.2 | NP_001010943.2 | 3 - 4 | 815 | 93 | |
XM_005633820.3 | XP_005633877.1 | 4 - 5 | 1192 | 93 | ||
XM_022408001.1 | XP_022263709.1 | 4 - 5 | 1195 | 93 | ||
GenBank mRNA | AF276427.1 | - | 3 - 4 | 815 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Canis familiaris Probe spans exons |