Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204843.1 | NP_001191772.1 | 4 - 5 | 320 | 90 | |
XM_022423974.1 | XP_022279682.1 | 4 - 5 | 482 | 90 | ||
GenBank mRNA | BM537539.1 | - | 4 - 5 | 324 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |