Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638401.3 | XP_005638458.1 | 2 - 3 | 596 | 61 | |
XM_022412503.1 | XP_022268211.1 | 1 - 2 | 106 | 61 | ||
XM_022412505.1 | XP_022268213.1 | 2 - 3 | 131 | 61 | ||
XM_022412506.1 | XP_022268214.1 | 1 - 2 | 106 | 61 | ||
XM_022412507.1 | XP_022268215.1 | 1 - 2 | 106 | 61 | ||
XM_022412508.1 | XP_022268216.1 | 1 - 2 | 106 | 61 | ||
XM_845774.5 | XP_850867.1 | 2 - 3 | 273 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |