Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846104.5 | XP_851197.1 | 3 - 4 | 437 | 108 | |
XM_858230.5 | XP_863323.1 | 3 - 4 | 427 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |