Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634758.3 | XP_005634815.1 | 1 - 2 | 333 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |