Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534762.6 | XP_534762.3 | 3 - 4 | 406 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |