Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_863139.5 | XP_868232.1 | 1 - 2 | 213 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |