Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405629.1 | XP_022261337.1 | 4 - 5 | 472 | 66 | |
XM_847452.5 | XP_852545.1 | 4 - 5 | 410 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |