Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC474791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_532021.6 | XP_532021.3 | 3 - 4 | 555 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |