Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616543.3 | XP_005616600.1 | 3 - 4 | 447 | 59 | |
XM_005616544.3 | XP_005616601.1 | 3 - 4 | 418 | 59 | ||
XM_533658.6 | XP_533658.1 | 3 - 4 | 479 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |