Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_532275.6 | XP_532275.2 | 2 - 3 | 276 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |