Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641610.2 | XP_005641667.1 | 2 - 3 | 221 | 105 | |
XM_005641611.2 | XP_005641668.1 | 2 - 3 | 217 | 105 | ||
XM_005641612.2 | XP_005641669.1 | 2 - 3 | 196 | 105 | ||
XM_538121.5 | XP_538121.1 | 2 - 3 | 230 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |