Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622749.3 | XP_005622806.1 | 3 - 4 | 368 | 80 | |
XM_005622751.3 | XP_005622808.1 | 3 - 4 | 367 | 80 | ||
XM_005622752.3 | XP_005622809.1 | 3 - 4 | 370 | 80 | ||
XM_537256.6 | XP_537256.2 | 3 - 4 | 365 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |