Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014116916.2 | XP_013972391.1 | 3 - 4 | 1325 | 65 | |
XM_014116919.2 | XP_013972394.1 | 3 - 4 | 1325 | 65 | ||
XM_022423534.1 | XP_022279242.1 | 3 - 4 | 1325 | 65 | ||
XM_022423535.1 | XP_022279243.1 | 3 - 4 | 1325 | 65 | ||
XM_022423537.1 | XP_022279245.1 | 3 - 4 | 1325 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |