Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014116841.2 | XP_013972316.1 | 1 - 2 | 162 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |