Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538701.6 | XP_538701.5 | 5 - 6 | 1089 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |