Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844226.4 | XP_849319.2 | 2 - 3 | 316 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |