Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639152.4 | XP_003639200.1 | 3 - 4 | 463 | 84 | |
XM_005622808.3 | XP_005622865.1 | 3 - 4 | 443 | 84 | ||
XM_022421392.1 | XP_022277100.1 | 3 - 4 | 503 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |