Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639517.3 | XP_005639574.1 | 2 - 3 | 279 | 79 | |
XM_005639518.2 | XP_005639575.1 | 2 - 3 | 228 | 79 | ||
XM_005639520.3 | XP_005639577.1 | 2 - 3 | 294 | 79 | ||
XM_005639521.2 | XP_005639578.1 | 2 - 3 | 638 | 79 | ||
XM_022414040.1 | XP_022269748.1 | 2 - 3 | 245 | 79 | ||
XM_022414041.1 | XP_022269749.1 | 2 - 3 | 304 | 79 | ||
XM_022414042.1 | XP_022269750.1 | 2 - 3 | 315 | 79 | ||
XM_022414043.1 | XP_022269751.1 | 2 - 3 | 318 | 79 | ||
XM_022414044.1 | XP_022269752.1 | 2 - 3 | 304 | 79 | ||
XM_022414045.1 | XP_022269753.1 | 2 - 3 | 188 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |