Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845717.5 | XP_850810.2 | 1 - 2 | 239 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |