Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847238.5 | XP_852331.3 | 1 - 2 | 482 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |