Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620592.2 | XP_005620649.1 | 550 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris |