Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637911.3 | XP_005637968.1 | 3 - 4 | 269 | 76 | |
XM_845222.5 | XP_850315.1 | 3 - 4 | 269 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |