Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF38B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022420382.1 | XP_022276090.1 | 5 - 6 | 783 | 100 | |
XM_849600.5 | XP_854693.1 | 3 - 4 | 579 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |