Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850300.5 | XP_855393.1 | 4 - 5 | 611 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |