Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022418186.1 | XP_022273894.1 | 591 | 85 | ||
XM_847911.4 | XP_853004.1 | 544 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |