Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623674.3 | XP_005623731.1 | 2 - 3 | 335 | 82 | |
XM_005623676.3 | XP_005623733.1 | 2 - 3 | 289 | 82 | ||
XM_022422231.1 | XP_022277939.1 | 2 - 3 | 251 | 82 |
Set Membership: |
![]() ![]() ![]() |