Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHURC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847755.5 | XP_852848.2 | 3 - 4 | 341 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |