Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022407261.1 | XP_022262969.1 | 3 - 4 | 206 | 75 | |
XM_849868.5 | XP_854961.1 | 3 - 4 | 273 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |