Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616534.3 | XP_005616591.1 | 2 - 3 | 714 | 93 | |
XM_850176.5 | XP_855269.1 | 2 - 3 | 750 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |