Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638782.4 | XP_003638830.1 | 355 | 82 | ||
XM_022418472.1 | XP_022274180.1 | 300 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |