Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635535.3 | XP_005635592.1 | 246 | 75 | ||
XM_534526.6 | XP_534526.2 | 246 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |