Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119564.2 | XP_013975039.1 | 6 - 7 | 2296 | 85 | |
XM_014119565.2 | XP_013975040.1 | 6 - 7 | 1831 | 85 | ||
XM_022427982.1 | XP_022283690.1 | 6 - 7 | 2207 | 85 | ||
XM_845204.5 | XP_850297.1 | 6 - 7 | 2206 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |