Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639628.3 | XP_003639676.1 | 2 - 3 | 267 | 61 | |
XM_003639630.3 | XP_003639678.1 | 2 - 3 | 336 | 61 | ||
XM_014120728.2 | XP_013976203.1 | 2 - 3 | 435 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |