Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630713.3 | XP_005630770.1 | 12 - 13 | 1286 | 170 | |
XM_022404930.1 | XP_022260638.1 | 11 - 12 | 1195 | 170 | ||
XM_022404931.1 | XP_022260639.1 | 11 - 12 | 1175 | 170 | ||
XM_022404932.1 | XP_022260640.1 | 11 - 12 | 1182 | 170 | ||
XM_844483.5 | XP_849576.1 | 12 - 13 | 1268 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Canis familiaris Probe spans exons |