Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617974.3 | XP_005618031.1 | 3 | 571 | 192 | |
XM_005617975.3 | XP_005618032.1 | 3 | 635 | 192 | ||
XM_014111400.2 | XP_013966875.1 | 3 | 503 | 192 | ||
XM_022414389.1 | XP_022270097.1 | 3 | 599 | 192 | ||
XM_022414391.1 | XP_022270099.1 | 4 | 709 | 192 | ||
XM_022414394.1 | XP_022270102.1 | 4 | 659 | 192 | ||
XM_843883.5 | XP_848976.1 | 3 | 585 | 192 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |