Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637831.3 | XP_005637888.1 | 4 - 5 | 531 | 83 | |
XM_022411750.1 | XP_022267458.1 | 4 - 5 | 531 | 83 | ||
XM_544060.5 | XP_544060.2 | 4 - 5 | 531 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |