Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620470.2 | XP_005620527.1 | 232 | 104 | ||
XM_844297.4 | XP_849390.2 | 232 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |