Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431974.4 | XP_003432022.1 | 2 - 3 | 266 | 81 | |
XM_014119445.2 | XP_013974920.1 | 2 - 3 | 280 | 81 | ||
XM_022427855.1 | XP_022283563.1 | 2 - 3 | 174 | 81 | ||
XM_022427856.1 | XP_022283564.1 | 2 - 3 | 166 | 81 | ||
XM_532565.6 | XP_532565.2 | 3 - 4 | 312 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |