Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410096.1 | XP_022265804.1 | 5 - 6 | 3129 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |