Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003362.1 | NP_001003362.1 | 7 - 8 | 1216 | 151 | |
XM_014106508.2 | XP_013961983.1 | 8 - 9 | 1392 | 151 | ||
XM_022405391.1 | XP_022261099.1 | 7 - 8 | 1216 | 151 | ||
XM_022405397.1 | XP_022261105.1 | 7 - 8 | 1216 | 151 | ||
XM_022405408.1 | XP_022261116.1 | 8 - 9 | 1233 | 151 | ||
XM_022405411.1 | XP_022261119.1 | 7 - 8 | 1216 | 151 | ||
XM_022405413.1 | XP_022261121.1 | 7 - 8 | 1216 | 151 | ||
XM_022405415.1 | XP_022261123.1 | 8 - 9 | 1349 | 151 | ||
XM_022405417.1 | XP_022261125.1 | 7 - 8 | 1216 | 151 | ||
XM_022405422.1 | XP_022261130.1 | 7 - 8 | 1216 | 151 | ||
XM_022405428.1 | XP_022261136.1 | 7 - 8 | 1216 | 151 | ||
XM_022405430.1 | XP_022261138.1 | 7 - 8 | 1216 | 151 | ||
XM_022405432.1 | XP_022261140.1 | 7 - 8 | 1216 | 151 | ||
XM_022405433.1 | XP_022261141.1 | 7 - 8 | 1216 | 151 | ||
GenBank mRNA | AY553843.1 | - | 7 - 8 | 1216 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |