Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_532463.6 | XP_532463.1 | 1 - 2 | 189 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |