Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118761.2 | XP_013974236.1 | 2 | 1631 | 161 | |
XM_022423625.1 | XP_022279333.1 | 2 | 1522 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |