Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639151.4 | XP_003639199.1 | 2 | 389 | 79 | |
XM_005622757.3 | XP_005622814.1 | 2 | 397 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |