Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845319.5 | XP_850412.1 | 2 | 758 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Inventoried Both primers and probe map within a single exon |