Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190429.2 | NP_001177358.2 | 2 | 387 | 113 | |
XM_005615772.3 | XP_005615829.1 | 3 | 593 | 113 | ||
GenBank mRNA | HM068511.1 | - | 2 | 388 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Both primers and probe map within a single exon |